(S)-(+)-2-Methylbutyric acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: (S)-(+)-2-Methylbutanoic acid, (S)-2-Methylbutanoic acid

245526-1G 245526-5G
SIAL245526-1GEA 158 GBP
SIAL245526-1G SIAL245526-5G
(S)-(+)-2-Methylbutyric acid, Sigma-Aldrich®
(S)-(+)-2-Methylbutyric acid
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 78 °C (1013 hPa)
Density: 0.9419 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00063166
CAS Number: 1730-91-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR