(S)-(+)-2-Amino-1-propanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: L(+)-Alaninol, L-Alaninol, (S)-2-Aminopropan-1-ol, L(+)-2-Amino-1-propanol

M-Clarity™ quality level = MQ100

A76206-10G A76206-1G
SIALA76206-10GEA 310 GBP
SIALA76206-10G SIALA76206-1G
(S)-(+)-2-Amino-1-propanol, Sigma-Aldrich®
(S)-(+)-2-Amino-1-propanol
Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 173
Density: 0.955 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064412
CAS Number: 2749-11-3
EINECS: 220-388-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR