L(+)-Glutamic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

G1251-100G G1251-1KG G1251-500G G1251-5KG
SIALG1251-100GEA 48.2 GBP
SIALG1251-100G SIALG1251-1KG SIALG1251-500G SIALG1251-5KG
L(+)-Glutamic acid, Sigma-Aldrich®
L(+)-Glutamic acid
Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.538 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002634
CAS Number: 56-86-0
EINECS: 200-293-7
Merck Index: 13,04482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR