L(+)-Aspartic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D
M-Clarity™ quality level = MQ300
W365601-10KG
W365601-1KG
W365601-SAMPLE
SIALW365601-10KGEA
512
GBP
SIALW365601-10KG
SIALW365601-1KG
SIALW365601-SAMPLE
L(+)-Aspartic acid, Sigma-Aldrich®
L(+)-Aspartic acid
Formula:
C₄H₇NO₄ MW: 133.1 g/mol Boiling Pt: 311 °C (1013 hPa) Melting Pt: 251 °C Density: 1.6613 g/cm³ (13 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002616 CAS Number: 56-84-8 EINECS: 200-291-6 Merck Index: 13,00845 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...