D-(-)-Cysteine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (S)-2-Amino-3-mercaptopropanoic acid, H-D-Cys-OH, (S)-(-)-Cysteine

30095-100MG 30095-1G
SIAL30095-100MGEA 75.9 GBP
SIAL30095-100MG SIAL30095-1G
D-(-)-Cysteine, Sigma-Aldrich®
D(-)-Cysteine
Formula: C₃H₇NO₂S
MW: 121.16 g/mol
MDL Number: MFCD00066461
CAS Number: 921-01-7
EINECS: 213-062-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR