L(+)-Cysteine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: H-Cys-OH, (R)-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid

M-Clarity™ quality level = MQ200

168149-100G 168149-2.5G 168149-25G 168149-500G
SIAL168149-100GEA 101 GBP
SIAL168149-100G SIAL168149-2.5G SIAL168149-25G SIAL168149-500G
L(+)-Cysteine, Sigma-Aldrich®
L(+)-Cysteine
Formula: HSCH₂CH(NH₂)CO₂H
MW: 121.16 g/mol
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.523 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064306
CAS Number: 52-90-4
EINECS: 200-158-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR