D-(-)-Arginine

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

4030317.0005 4030317.0025
BACE4030317.0005EA 0 GBP
BACE4030317.0005 BACE4030317.0025
D-(-)-Arginine
D(-)-Arginine
Formula: C₆H₁₄N₄O₂
Melting Pt: 212 °C
MDL Number: MFCD00063116
CAS Number: 157-06-2
EINECS: 205-866-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR