Ruthenium(III) chloride hydrate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Ruthenium(III) chloride xhydrate, Ruthenium trichloride hydrate

005538-1G 005538-5G 005538-25G
FLUO005538-1GEA 0 GBP
FLUO005538-1G FLUO005538-5G FLUO005538-25G
Ruthenium(III) chloride hydrate
Ruthenium (III) chloride hydrate
Formula: Cl₃Ru·xH₂O
Melting Pt: > 300 °C
Density: 3.11 g/cm³ (20 °C)
MDL Number: MFCD00149844
CAS Number: 14898-67-0
EINECS: 233-167-5
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR