Manganese(II) sulphate hydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
10807.06
10807.14
10807.06EA
41.9
GBP
10807.06
10807.14
Manganese(II) sulphate hydrate ≥99.999% (metals basis), Puratronic®
Manganese (II) sulphate hydrate
Purity: 99.999%
CAS No.: 15244-36-7
Molecular Formula: MnSO4.xH2O
Formula Weight: 151.00(anhy)
Physical Form: Crystalline Powder
Sensitivity: Hygroscopic
pH: 3-3.5
Merck Reference: 14,5739
UN No.: 3077
MDL No.: MFCD00149159
Formula:
MnO₄S·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149159 CAS Number: 15244-36-7 EINECS: 232-089-9 UN: 3077 ADR: 9,III Merck Index: 14,5739 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...