Zinc sulphate heptahydrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ200

Z0635-100G Z0635-500G
SIALZ0635-100GEA 89.6 GBP
SIALZ0635-100G SIALZ0635-500G
Zinc sulphate heptahydrate, Sigma-Aldrich®
Zinc sulphate heptahydrate
Formula: ZnSO₄·7H₂O
Melting Pt: 100 °C
Density: 1.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149894
CAS Number: 7446-20-0
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Merck Index: 12,10293

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR