Cerium trinitrate hexahydrate ≥99.9% (Ce basis)

Supplier: Strem Chemicals, Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Cerous chloride hexahydrate, Cerium (III) nitrate hexahydrate

93-5831-500G AB117425.0100
STRE93-5831-500GEA 276.21 GBP
STRE93-5831-500G ABCRAB117425.0100
Cerium trinitrate hexahydrate ≥99.9% (Ce basis)
Cerium trinitrate hexahydrate
Formula: Ce(NO₃)₃·6H₂O
MW: 434.22 g/mol
Boiling Pt: 200
Melting Pt: 39
Density: 2.377 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149631
CAS Number: 10294-41-4
EINECS: 233-297-2
UN: 1477
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR