Cerium trichloride heptahydrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Cerous chloride heptahydrate, Cerium (III) chloride heptahydrate

22300-100G 22300-500G
SIAL22300-100GEA 156 GBP
SIAL22300-100G SIAL22300-500G
Cerium trichloride heptahydrate, Sigma-Aldrich®
Cerium trichloride heptahydrate
Formula: CeCl₃·7H₂O
MW: 372.58 g/mol
Melting Pt: 848 °C
Density: 3.97 g/cm³ (20 °C)
MDL Number: MFCD00149634
CAS Number: 18618-55-8
EINECS: 232-227-8
Merck Index: 12,02041

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR