EDTA tetrasodium salt hydrate, Sigma-Aldrich®
Supplier: MerckTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Tetrasodium ethylenediaminetetraacetate hydrate, Kestranal® 4S hydrate, Ethylenedinitrilotetra-acetic acid tetrasodium salt hydrate, Sodium 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate xhydrate, Edetic acid tetrasodium salt hydrate, Ethylenediaminetetraacetic acid tetrasodium salt hydrate
M-Clarity™ quality level = MQ200
Formula:
C₁₀H₁₂N₂O₈Na₄·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150025 CAS Number: 13235-36-4 EINECS: 200-573-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...