Guanidinium thiocyanate

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

24220.02 24220.03
SERA24220.02EA 0 GBP
SERA24220.02 SERA24220.03
Guanidinium thiocyanate
Guanidinium thiocyanate
Formula: CH₅N₃·CHNS
MW: 118.16 g/mol
Melting Pt: 118 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013027
CAS Number: 593-84-0
EINECS: 209-812-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR