Hydrochloric acid 10%
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Hydrochloric acid solution, Muriatic acid
0710.1
0710.2
0710.3
ROTH0710.1EA
0
GBP
ROTH0710.1
ROTH0710.2
ROTH0710.3
Hydrochloric acid 10%
Hydrochloric acid
Formula:
HCl Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...