Trimethylamine 25% (w/w) in aqueous solution, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ300

W324108-10KG-K W324108-1KG-K W324108-20KG-K W324108-5KG-K
SIALW324108-10KG-KEA 333 GBP
SIALW324108-10KG-K SIALW324108-1KG-K SIALW324108-20KG-K SIALW324108-5KG-K
Trimethylamine 25% (w/w) in aqueous solution, Sigma-Aldrich®
Trimethylamine
Formula: C₃H₉N
MW: 59.11 g/mol
Boiling Pt: 2.87
Melting Pt: –3 °C
Density: 0.85…0.9 g/cm³ (20 °C)
Flash Pt: -20
Storage Temperature: Ambient
MDL Number: MFCD00008327
CAS Number: 75-50-3
EINECS: 200-875-0
UN: 1297
ADR: 3 II
Merck Index: 12,09841

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR