Hydrochloric acid 1 mol/l (1 N)

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

K025.1 K025.2
ROTHK025.1EA 20.84 GBP
ROTHK025.1 ROTHK025.2
Hydrochloric acid 1 mol/l (1 N)
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR