Acetic acid 60%
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Vinegar acid, Methanecarboxylic acid, Ethanoic acid
7329.1
7329.2
ROTH7329.1EA
0
GBP
ROTH7329.1
ROTH7329.2
Acetic acid 60%
Acetic acid
Formula:
H₃CCOOH Storage Temperature: Ambient |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 EINECS: 200-580-7 UN: 2790 ADR: 8,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...