Hydrochloric acid 0.1 mol/l (0.1 N)
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Hydrochloric acid solution, Muriatic acid
K024.1
K024.2
ROTHK024.1EA
22.18
GBP
ROTHK024.1
ROTHK024.2
Hydrochloric acid 0.1 mol/l (0.1 N)
Hydrochloric acid
Formula:
HCl Density: 1.004 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...