Glutaraldehyde 25% in aqueous solution

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial

23114.01 23114.02
SERA23114.01EA 28.06 GBP
SERA23114.01 SERA23114.02
Glutaraldehyde 25% in aqueous solution
Glutaraldehyde
Formula: C₅H₈O₂
Boiling Pt: 101 °C (1013 hPa)
Melting Pt: –6 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: >71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007025
CAS Number: 111-30-8
EINECS: 203-856-5
UN: 2927
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR