(±)-1-Penten-3-ol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (±)-Ethyl vinyl carbinol, Pent-1-en-3-ol

M-Clarity™ quality level = MQ300

W358401-100G-K W358401-1KG-K
SIALW358401-100G-KEA 159 GBP
SIALW358401-100G-K SIALW358401-1KG-K
(±)-1-Penten-3-ol, Sigma-Aldrich®
(±)-1-Penten-3-ol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 114 °C (1013 hPa)
Density: 0.838 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004573
CAS Number: 616-25-1
EINECS: 210-472-1
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR