DL-Tyrosine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Hydroxyphenylalanine, 3-(p-Hidroxifenil)-DL-alanina, H-DL-Tyr-OH

M-Clarity™ quality level = MQ100

145726-100G 145726-25G
SIAL145726-100GEA 316 GBP
SIAL145726-100G SIAL145726-25G
DL-Tyrosine, Sigma-Aldrich®
DL-Tyrosine
Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: 285 °C
MDL Number: MFCD00063074
CAS Number: 556-03-6
EINECS: 209-113-1
Merck Index: 12,09970

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR