(±)-3-Methylvaleric acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
3-Methylvaleric Acid, 3-Methylpentanoic acid, (±)-3-Methylpentanoic acid
002768-25G
002768-5G
FLUO002768-25GEA
0
GBP
FLUO002768-25G
FLUO002768-5G
(±)-3-Methylvaleric acid
(±)-3-Methylvaleric acid
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 196 °C (1013 hPa) Melting Pt: –41 °C Density: 0.93 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002729 CAS Number: 105-43-1 EINECS: 203-297-7 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...