(±)-2-Methylbutyric acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(±)-2-Methylbutanoic acid, 2-Methylbutyric acid, 2-Methylbutanoic acid
M-Clarity™ quality level = MQ300
W269506-1KG-K
W269506-25KG-K
W269506-5KG-K
SIALW269506-1KG-KEA
155
GBP
SIALW269506-1KG-K
SIALW269506-25KG-K
SIALW269506-5KG-K
(±)-2-Methylbutyric acid, Sigma-Aldrich®
(±)-2-Methylbutyric acid
Formula:
C₅H₁₀O₂ MW: 102.13 g/mol Boiling Pt: 177 °C (1013 hPa) Melting Pt: –70 °C Density: 0.938 g/cm³ (20 °C) Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002669 CAS Number: 116-53-0 EINECS: 204-145-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...