(±)-2-Methylbutyric acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (±)-2-Methylbutanoic acid, 2-Methylbutyric acid, 2-Methylbutanoic acid

M-Clarity™ quality level = MQ100

193070-100G 193070-25G
SIAL193070-100GEA 48.2 GBP
SIAL193070-100G SIAL193070-25G
(±)-2-Methylbutyric acid, Sigma-Aldrich®
(±)-2-Methylbutyric acid
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 177 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.938 g/cm³ (20 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00002669
CAS Number: 116-53-0
EINECS: 204-145-2
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR