Cyclohexene stabilised, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
29240-2.5L
29240-1L
29240-500ML
SIAL29240-2.5LEA
193
GBP
SIAL29240-2.5L
SIAL29240-1L
SIAL29240-500ML
Cyclohexene stabilised, Sigma-Aldrich®
Cyclohexene
Formula:
C₆H₁₀ MW: 82.15 g/mol Boiling Pt: 83 °C (1013 hPa) Melting Pt: –104 °C Density: 0.81 g/cm³ (20 °C) Flash Pt: –20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001539 CAS Number: 110-83-8 EINECS: 203-807-8 UN: 2256 ADR: 3,II Merck Index: 13,02756 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...