L(+)-Sodium lactate ≥99.0% (by titrimetric analysis), crystalline research grade

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

29763.01 29763.02
SERA29763.01EA 0 GBP
SERA29763.01 SERA29763.02
L(+)-Sodium lactate ≥99.0% (by titrimetric analysis), crystalline research grade
L(+)-Sodium lactate
Formula: C₃H₅NaO₃
Storage Temperature: Ambient
MDL Number: MFCD00066576
CAS Number: 867-56-1
EINECS: 200-772-0
Merck Index: 13,08709

Order Now

Specification Test Results

Assay (titrimetric) 99.0% min.
Solubility (1% solution in H₂O) Clear and colorless
[α]20D (1% solution in H₂O) -12.0° ± 1.0°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR