Tannic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Gallotannin, Tannin
M-Clarity™ quality level = MQ200
403040-100G
403040-500G
403040-50G
SIAL403040-100GEA
42.3
GBP
SIAL403040-100G
SIAL403040-500G
SIAL403040-50G
Tannic acid, Sigma-Aldrich®
Tannic acid
Formula:
C₇₆H₅₂O₄₆ Melting Pt: 218…219 °C Density: >1 g/cm³ (20 °C) Flash Pt: 199 °C |
MDL Number:
MFCD00066397 CAS Number: 1401-55-4 EINECS: 215-753-2 Merck Index: 12,09221 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...