Tween® 80 (Polysorbate)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EMR245001
EMR245005
EMR245025
EURLEMR245001EA
0
GBP
EURLEMR245001
EURLEMR245005
EURLEMR245025
Tween® 80 (Polysorbate)
Tween® 80 (Polysorbate)
Boiling Pt:
∼100 °C (1013 hPa) Melting Pt: –21 °C Density: 1.062 g/cm³ (20 °C) Flash Pt: >149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082107 CAS Number: 9005-65-6 EINECS: 500-019-9 Merck Index: 13,07664 |