Polyethylene glycol 6,000

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: PEG 6000PEG 6,000

SERA33137.01EA 16.87 GBP
SERA33137.01 SERA33137.02
Polyethylene glycol 6,000
Polyethylene glycol 6,000
Formula: H(OCH₂CH₂)nOH
MW: 6000 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 55…62 °C
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01779614
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR