Polyethylene glycol 4,000
Supplier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
33136.01
33136.02
SERA33136.01EA
27.83
GBP
SERA33136.01
SERA33136.02
Polyethylene glycol 4,000
Polyethylene glycol 4,000
Formula:
H(OCH₂CH₂)nOH Boiling Pt: ∼250 °C (1013 hPa) Melting Pt: 53…58 °C Density: 1.13 g/cm³ (20 °C) Flash Pt: 250 °C Storage Temperature: Ambient |
MDL Number:
MFCD01779612 CAS Number: 25322-68-3 EINECS: 500-038-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...