Human albumin (from serum)

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

47323.02 47323.03
SERA47323.02EA 0 GBP
SERA47323.02 SERA47323.03
Human albumin (from serum)
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name Albumin
Environmentally Preferable
Protein synonyms Human Serum Albumin|HSA|serum albumin|HAS|serum of albumin
Species Human
Source Serum
CAS No. 70024-90-7
Storage conditions Refrigerator

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR