Gum arabic, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

51198-1KG 51198-250G
SIAL51198-1KGEA 363 GBP
SIAL51198-1KG SIAL51198-250G
Gum arabic, Sigma-Aldrich®
Gum arabic
Density: 1.35 g/cm³ (20 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00081264
CAS Number: 9000-01-5
EINECS: 232-519-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR