D-(+)-Xylose

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

38530.01 38530.02
SERA38530.01EA 0 GBP
SERA38530.01 SERA38530.02
D-(+)-Xylose
D(+)-Xylose
Formula: C₅H₁₀O₅
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 154 °C
Density: 1.525 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151475
CAS Number: 58-86-6
EINECS: 200-400-7
Merck Index: 13,10142
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f045c9c134fcd7d287b95738095c40d429a173e&vid=447bd80bb2f3ca1cb52dc711d0957e9ab6e2f84d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR