D-(+)-Galactose, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
M-Clarity™ quality level = MQ200
15522-250G-R
15522-100G-R
SIAL15522-250G-REA
218
GBP
SIAL15522-250G-R
SIAL15522-100G-R
D-(+)-Galactose, Sigma-Aldrich®
D(+)-Galactose
Formula:
C₆H₁₂O₆ Boiling Pt: 527 °C (1013 hPa) Melting Pt: 163…169 °C Density: 1.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00151230 CAS Number: 59-23-4 EINECS: 200-416-4 Merck Index: 13,04356 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...