Benzene-[D₆], Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Benzene-[²H₆]
M-Clarity™ quality level = MQ100
175978-50G
SIAL175978-50GEA
319
GBP
SIAL175978-50G
Benzene-[D₆], Sigma-Aldrich®
Benzene-[D6]
Formula:
C₆D₆ MW: 84.07 g/mol Boiling Pt: 79 °C (1013 hPa) Melting Pt: 6.7 °C Density: 0.95 g/cm³ (20 °C) Flash Pt: –11 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00003010 CAS Number: 1076-43-3 EINECS: 214-061-8 UN: 1114 ADR: 3,II |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...