Isoeugenol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Propenylguaiacol4-Hydroxy-3-methoxy-1-propenylbenzene2-Methoxy-4-(prop-1-en-1-yl)phenol

M-Clarity™ quality level = MQ100

SIALI17206-100GEA 87.2 GBP
SIALI17206-100G SIALI17206-5G
Isoeugenol, Sigma-Aldrich®
Isoeugenol
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: –10 °C
Density: 1.09 g/cm³ (20 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00009285
CAS Number: 97-54-1
EINECS: 202-590-7
Merck Index: 13,05189

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR