Palladium ≥99.99% (trace metals basis), granules, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

203998-1G 203998-5G
SIAL203998-1GEA 446 GBP
SIAL203998-1G SIAL203998-5G
Palladium ≥99.99% (trace metals basis), granules, Sigma-Aldrich®
Palladium
Formula: Pd
MW: 106.420 g/mol
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR