Molybdenum, gauze woven from wire 35 mesh, Ø 0.0509 mm (0.002 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

42485.FL 42485.GT 42485.GV
42485.FLEA 0 GBP
42485.FL 42485.GT 42485.GV
Molybdenum, gauze woven from wire 35 mesh, Ø 0.0509 mm (0.002 in)
Molybdenum

CAS No.: 7439-98-7 Physical Form: Wire Cloth MDL No.: MFCD00003465

Formula: Mo
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR