Bismuth, powder, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

265462-100G 265462-500G
SIAL265462-100GEA 91.2 GBP
SIAL265462-100G SIAL265462-500G
Bismuth, powder, Sigma-Aldrich®
Bismuth
Formula: Bi
MW: 208.98 g/mol
Boiling Pt: 1490
Density: 9.78 g/cm³ (20 °C)
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,01297

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR