Tin, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
217697-100G
217697-500G
SIAL217697-100GEA
81.9
GBP
SIAL217697-100G
SIAL217697-500G
Tin, Sigma-Aldrich®
Tin
Formula:
Sn MW: 118.71 g/mol Boiling Pt: 2270 Melting Pt: 232 Density: 7.31 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...