Charcoal activated, powder, puriss. p.a., Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Carbon activated, Activated carbon, Activated charcoal

M-Clarity™ quality level = MQ100

31616-250G 31616-750G
SIAL31616-250GEA 49.4 GBP
SIAL31616-250G SIAL31616-750G
Charcoal activated, powder, puriss. p.a., Supelco®
Charcoal activated
Formula: C
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR