Germanium, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
203343-25G
203343-5G
SIAL203343-25GEA
283
GBP
SIAL203343-25G
SIAL203343-5G
Germanium, Sigma-Aldrich®
Germanium
Formula:
Ge MW: 72.61 g/mol Boiling Pt: 2830 °C (1013 hPa) Melting Pt: 937.2 °C Density: 5.323 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085310 CAS Number: 7440-56-4 EINECS: 231-164-3 Merck Index: 12,04415 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...