Boron, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ100
266620-25G
266620-5G
SIAL266620-25GEA
678
GBP
SIAL266620-25G
SIAL266620-5G
Boron, Sigma-Aldrich®
Boron
Formula:
B Density: 1.73 g/cm³ (15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134034 CAS Number: 7440-42-8 EINECS: 231-151-2 Merck Index: 13,01333 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...