Calcium, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ100
215147-100G
215147-500G
215147-5G
SIAL215147-100GEA
152
GBP
SIAL215147-100G
SIAL215147-500G
SIAL215147-5G
Calcium, Sigma-Aldrich®
Calcium
Formula:
Ca Boiling Pt: 1484 °C (1013 hPa) Melting Pt: 850 °C Density: 1.54 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085314 CAS Number: 7440-70-2 EINECS: 231-179-5 UN: 1401 ADR: 4.3,II Merck Index: 13,01644 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...