General purpose ovens
Supplier: GENLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OV/125/CLAD OV100/CLAD/F/D OV125/CLAD/F/D OV150/CLAD/F/D OV200/CLAD/F/D OV/50/TDIG/F OV75/TDIG/F OV50DIG
GENLOV/125/CLADEA 0 GBP
GENLOV/125/CLAD 466-0239 466-0240 466-0241 466-0242 466-0243 466-0244 GENLOV50DIG
General purpose ovens
Ovens Standard Ovens
A range of efficient, reliable, cost effective ovens ideal for most drying, warming and general laboratory applications. The exterior is sheet steel finished in an easy-to-clean powder coated paint and the inner chamber is made from mild steel coated with aluminium (CLAD). The top vent is fitted with a clip to hold a mercury-in-glass thermometer. Heated by means of Incoloy sheathed elements; positioned below the chamber floor for natural convection units and fitted around the fan on the back or side wall of the chamber for mechanical convection units. The control system comprises of a directMore Product Information