Fmoc-His(pi-Me)-OH ≥98.0% (by HPLC)

Supplier: Novabiochem (Part of Merck)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Fmoc-His(3-Me)-OH, Fmoc-3-methyl-L-histidine

8.52286.8250 8.52286.0001
8.52286.8250EA 142 GBP
8.52286.8250 8.52286.0001
Fmoc-His(pi-Me)-OH ≥98.0% (by HPLC)
Fmoc-His(pi-Me)-OH

Building block for the incorporation of 1-methylhistidine by Fmoc SPPS.

Formula: C₂₂H₂₁N₃O₄
MW: 391.427 g/mol
MDL Number: MFCD02259494
CAS Number: 252049-16-4

Order Now

Specification Test Results

Colour (visual) white to slight yellow to beige
Appearance of substance (visual) Powder
Identity (IR) Passes test
Assay (HPLC, area%) ≥98,0%
Solubility (0,5 mmol in 4 ml NMP + 500 µl DIPEA) Clearly soluble
Storage temperature 2 − 8 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR