Sample bottles, with label
Supplier: Local
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LOCAQ1EA 110 GBP
LOCAQ1 LOCAR2 LOCAR1 LOCAR4 LOCAP2 LOCAQ3 LOCAP1 LOCAQ2 LOCAR3 LOCAP4 LOCAQ5 LOCAQ4 LOCAP3 LOCAR5 LOCAP5
Sample bottles, with label
Bottles Plastic Bottles
Plastic, translucent, with screw cap.
These Cribbar sample bottles are sodium thiosulphate dosed or EDTA dosed or undosed, available as sterile and unsterile formats.