9(R)-HODE 0,01% in ethanol , 98%

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 9R-hydroxy-10E,12Z-octadecadienoic acid

38405-25 38405-50 38405-100 38405-250
CAYM38405-25EA 203 GBP
CAYM38405-25 CAYM38405-50 CAYM38405-100 CAYM38405-250
9(R)-HODE 0,01% in ethanol , 98%
9(R)-HODE

9(R)-HODE is one of several monohydroxylated products of linoleic acid. All known mammalian lipoxygenases appear to catalyze the oxygenation of arachidonic and linoleic acid to give products having strictly the (S) configuration at the site of oxygen insertion. However, both human umbilical vein endothelial cells and bovine aorta endothelial cells have been shown to produce 9(R)-HODE when incubated with linoleic acid.
The physiological function of 9(R)-HODE and the enzyme that catalyzes its formation have not been determined.


  • Purity: ≥98%
  • Formulation: A solution in ethanol
  • λmax: 234 nm


Warning - this product is not for human or veterinary use.

Formula: C₁₈H₃₂O₃
MW: 296.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00084832
CAS Number: 10075-11-3
UN: 1170
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR