Performance oil No.19, for pumps, ULTRAGRADE™
Supplier: EDWARDS HIGH VACUUM METTLER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H110-25-012 H110-25-011
181-0225EA 53.4 GBP
181-0225 181-0224
Performance oil No.19, for pumps, ULTRAGRADE™
Lubricants
ULTRAGRADE™ Performance 19 oil manufactured by a unique hydrotreating process instead of solvent refining process. This process eliminates nitrogen, sulphur and oxygen and the aromatic hydrocarbons, leaving a clear base stock essentially free of impurities.
  • Ideal for large pumps used in heavy-duty, cycling applications
  • Extending intervals between oil changes
The combination of this oil with additives give improved vacuum performance, thermal and oxidation stability and rust inhibition. The anti-oxidants present in the oil helps the pumps to withstandMore Product Information